Methanol ≥99.9%, ULTRA RESI-ANALYZED® for gas chromatography, for purge and trap GC analysis, J.T.Baker®

Supplier: Avantor
Danger


Packaged under inert gas.

BAKR9077-02EA 354 SGD
BAKR9077-02
Methanol ≥99.9%, ULTRA RESI-ANALYZED® for gas chromatography, for purge and trap GC analysis, J.T.Baker®
Methanol
Formula: CH₃OH
Boiling Pt: 64,6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0,7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 99.9 %
Assay (CH₃OH) (by GC, corrected for water) ≥99.9 %
Residue after Evaporation ≤1.0 ppm
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.10
Water (by KF, coulometric) ≤0.08 %
Volatile Organic Trace Analysis - Below EPA 8260B CRQL Conforms / Fails
Performance Tested for Use in EPA Methods
500 Series for Drinking Water
600 Series for Wastewater
846 for Solid Waste


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR