Methanol ≥99.9% (by GC, corrected for water content) for HPLC, for spectrophotometry, J.T.Baker®

Supplier: Avantor
Danger


Filtered through a 0.2 µm filter.

BAKR9124-80EA 26 SGD
BAKR9124-80
Methanol ≥99.9% (by GC, corrected for water content) for HPLC, for spectrophotometry, J.T.Baker®
Methanol
Formula: CH₃OH
Boiling Pt: 64,6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0,7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CH₃OH) (by GC, corrected for water) ≥99.9 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 225 nm ≤0.15
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤205
Gradient Elution Test (a.u.) - 254 nm ≤0.002
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Acetone ≤0.001 %
Residue after Evaporation ≤1.0 ppm
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.10
Water (by KF, coulometric) ≤0.05 %
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR