Acetonitrile for HPLC, for spectrophotometry, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

BAKR9152-80EA 81.7 SGD
BAKR9152-80
Acetonitrile for HPLC, for spectrophotometry, J.T.Baker®
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81,6 °C (1013 hPa)
Melting Pt: –45,7 °C
Density: 0,782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 200 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤190
Gradient Elution Test (a.u.) - 254 nm ≤0.005
Gradient Elution Test (a.u.) - 210 nm ≤0.005
Density (g/mL) at 25°C 0.775 - 0.780
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Assay (CH₃CN) (by GC) ≥99.9 %
Appearance Passes Test
Color (APHA) ≤10
Residue after Evaporation ≤1.0 ppm
Titrable Acid (µeq/g) ≤0.8
Titrable Base (µeq/g) ≤0.6
Water (by KF, coulometric) ≤100 ppm
Carbonyl Compounds (as Acetone) ≤25.0 ppb
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR