ALCH, anhydrous 94.0-96.0% (v/v), PHOTREX® for spectrophotometry, for liquid chromatography (denatured with 5% isopropanol and 5% methanol), J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Reagent alcohol

BAKR9229-03EA 113 SGD
BAKR9229-03 BAKR9229-01
ALCH, anhydrous 94.0-96.0% (v/v), PHOTREX® for spectrophotometry, for liquid chromatography (denatured with 5% isopropanol and 5% methanol), J.T.Baker®
Alcohol

Made from Specially Denatured Alcohol 3A.


  • Total alcohol: ~100%
  • Ethyl alcohol: 89 to 92%
  • Denaturant: 4 to 6% methyl alcohol and 4 to 6% 2-propanol


For Use in Liquid Chromatography and Spectrophotometry. Ultraviolet Absorbance (1,00-cm cell vs. water) curve smooth throughout stated range with no extraneous impurity peaks.


SDA Formula 3A consists of 5 volumes of methanol and 100 volumes of 200 proof ethanol.

Boiling Pt: 78,3 °C (1013 hPa)
Melting Pt: –114 °C
Density: 0,7895 g/cm³ (20 °C)
Flash Pt: 13 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1987
ADR: 3,II
REACH: 01-2119457610-43
Merck Index: 12,03806

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 94.0 - 96.0 %
Assay Specially Denatured Alcohol Formula 3A (200 proof)(v/v) 94.0 - 96.0 %
Isopropyl Alcohol (2-propanol)(v/v) 4.0 - 6.0 %
Ethanol (v/v) 88.5 - 92.5 %
Methanol (v/v) 3.5 - 5.5 %
Appearance Passes Test
Color (APHA) ≤5
Residue after Evaporation ≤5 ppm
Specific Gravity at 25°C 0.785 - 0.795
Water (H₂O) ≤0.1 %
Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 210 nm ≤1.00
Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 220 nm ≤0.50
Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 230 nm ≤0.20
Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 250 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 270 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 400 nm ≤0.01
Substances Reducing Permanganate Passes Test
Titrable Acid (µeq/g)
Titrable Base (µeq/g)
SDA Formula 3A consists of 5 volumes of methanol and 100 volumes of 200proof ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR