Hexane (mixture of isomers) ≥99.5% (by GC, corrected for water content) (n-hexane ≥95%), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Hexanes, Isohexane

Packaged under inert gas.

BAKR9262-03EA 270 SGD
BAKR9262-03 BAKR9262-02
Hexane (mixture of isomers) ≥99.5% (by GC, corrected for water content) (n-hexane ≥95%), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94,3 °C
Density: 0,659 g/cm³ (20 °C)
Flash Pt: –23 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 5
FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL) ≤ 5
ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL) ≤ 10
ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL) ≤ 5
Assay (Total Saturated C₆ Isomers) (by GC, corrected for water) ≥ 99.5 %
Assay (as n-Hexane) (by GC, corrected for water) ≥ 95 %
Color (APHA) ≤ 10
Residue after Evaporation ≤ 1.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤ 0.05 %
MEETS SPECIFICATIONS WITHIN THE EXPIRATION PERIOD


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR