Hexane (mixture of isomers) 98,5 GC, sum of enantiomers, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Hexanes, Isohexane

BAKR9309-05EA 414 SGD
BAKR9309-05 BAKR9309-07 BAKR9309-22 BAKR9309-03 BAKR9309-R BAKR9309-01 BAKR9309-33
Hexane (mixture of isomers) 98,5 GC, sum of enantiomers, BAKER ANALYZED® ACS, J.T.Baker®
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.663 g/cm³ (25 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 98.5 %
ACS - Assay (Sum of 5 isomers, total hexanes plus methylcyclopentane) (by GC) ≥ 98.5 %
ACS - Color (APHA) ≤ 10
ACS - Residue after Evaporation ≤ 0.001 %
ACS - Water-Soluble Titrable Acid, meq/g ≤ 0.0003
ACS - Sulfur Compounds (as S) ≤ 0.005 %
ACS - Thiophene Passes Test
Product Information (not specifications): 68.7 °C
Boiling Point (typical)................. 68.7 °C. 0.663
Density (g/mL) at 25 °C (typical)........ 0.663

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR