Methyl ethyl ketone ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

Management of Change (MOC) category = N

BAKR9319-22EA 0 SGD
BAKR9319-22
Methyl ethyl ketone ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Methyl ethyl ketone

The quality of many BAKER ANALYZED™ Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These solvents are essential for any chemical stockroom.

Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6
Melting Pt: –86 °C
Density: 0.8054 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3 II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use = 99.0 %
Assay (CH₃COCH₂CH₃) (by GC, corrected for water) ≥99.0 %
Color (APHA) ≤15
Residue after Evaporation ≤0.001 %
Titrable Acid (µeq/g) ≤0.5
Water (by KF, volumetric) ≤0.05 %
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR