ALCH, anhydrous 94,0-96,0 v/v, BAKER ANALYZED® ACS (MeOH+ IPA), J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Reagent alcohol

BAKR9401-06EA 74.9 SGD
BAKR9401-06 BAKR9401-22 BAKR9401-03 BAKR9401-07 BAKR9401-05 BAKR9401-02
ALCH, anhydrous 94,0-96,0 v/v, BAKER ANALYZED® ACS (MeOH+ IPA), J.T.Baker®
Alcohol

Alcohol, anhydrous, reagent made from specially denatured alcohol 3A.


  • Total alcohol: 99,5%
  • Ethyl alcohol: 89 to 92%
  • Denaturant: 2,5 to 10% methyl alcohol and 4 to 6% 2-propanol

Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1987
ADR: 3 II

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements 94.0 - 96.0 %
For Laboratory,Research,or Manufacturing Use 4.0 - 6.0 %
Assay Specially Denatured Alcohol Formula 3A (200 proof)(v/v) 94.0 - 96.0 %
Isopropyl Alcohol (2-propanol)(v/v) 4.0 - 6.0 %
Appearance Passes Test
Color (APHA) ≤ 10
Residue after Evaporation ≤ 0.001 %
Specific Gravity at 25°C 0.785 - 0.795
Water (H₂O) ≤ 0.5 %
SDA Formula 3A consists of 5 volumes of methanol and 100 volumes of 200proof ethanol.
NOTE: Intended for nonfuel end purposes only.
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR