Hydrochloric acid 37%, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

BAKR9553-69EA 7530 SGD
BAKR9553-69
Hydrochloric acid 37%, J.T.Baker®
Hydrochloric acid
Formula: HCl
MW: 36.46
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR