N,N-Dimethylacetamide, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®

Supplier: Avantor

BAKR9755.1000EA 134 SGD
BAKR9755.1000
N,N-Dimethylacetamide, BAKER ANALYZED® for headspace gas chromatography, J.T.Baker®
N,N-Dimethylacetamide
Formula: CH₃CON(CH₃)₂
MDL Number: MFCD00008686
CAS Number: 127-19-5
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace04c31b8db2d1fc0c18051cd2eefc52a61ea65&vid=4e98bd4c34a9326a56ee3e11797b2e6e1b323c8f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) min. 99.9 %
Acidity (meq/g) max. 0.0005
Evaporation Residue max. 0.0005 %
GC-HS Suitability passes test
Water (KF) max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR