J.T.Baker®, Liquid Level Indicators
Supplier: Avantor
BAKR9803-03EA 2670 SGD
BAKR9803-03
J.T.Baker®, Liquid Level Indicators
Level Sensors
These level indicators provide the digital readout of the approximate volume of solvent remaining in the CYCLE-TAINER system.
  • Signal can also be used to drive switching circuits