J.T.Baker®, Cycle-Tap Sampler
Supplier: Avantor
BAKR9815-05EA 105 SGD
BAKR9815-05
J.T.Baker®, Cycle-Tap Sampler
Samplers Fluid Samplers
Cycle-Tap sampler is a device for sampling small quantities of solvent.
Designed to eliminate solvent exposure to the atmosphere when used with a Luer lock syringe.