Acetonitrile, BAKER ANALYZED® for LC-MS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

BAKR9821.2500GLEA 78.4 SGD
BAKR9821.2500GL
Acetonitrile, BAKER ANALYZED® for LC-MS, J.T.Baker®
Acetonitrile
Formula: H₃CCN
Boiling Pt: 81,6 °C (1013 hPa)
Melting Pt: –45,7 °C
Density: 0,782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (by GC) min. 99.9 %
Residue after Evaporation max. 1 ppm
Water (H2O) max. 0.01 %
LC-Gradient-Diode Array Detection (AU), test solution is modified with 0.1% (v/v) formic acid:
at 220 nm max. 0.01
at 254 nm max. 0.01
LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), test solution is modified with 0.1% (v/v) formic acid:
Positive ESI-MS Sensitive Impurities (as Reserpine) max. 50 ng/ml
Trace Impurities (in ppb)
Aluminium (Al) max. 50
Calcium (Ca) max. 50
Iron (Fe) max. 50
Potassium (K) max. 50
Magnesium (Mg) max. 50
Sodium (Na) max. 50
Ultraviolet Absorbance (1.00-cm path vs water):
at 200 nm max. 0.05
Element concentrations are at time of lot release.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR