Water, BAKER ANALYZED®, J.T.Baker®

Supplier: Avantor

BAKR9825.2500GLEA 54.9 SGD
BAKR9825.2500GL
Water, BAKER ANALYZED®, J.T.Baker®
Water
Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Residue after Evaporation max. 5 ppm
LC-Gradient-Diode Array Detection (AU), test solution is modified with 0.1% (v/v) formic acid:
at 210 nm max. 0.005
at 254 nm max. 0.001
LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), test solution is modified with 0.1% (v/v) formic acid:
Positive ESI-MS Sensitive Impurities (as Reserpine) max. 100 ng/ml
Trace Impurities (in ppb)
Aluminium (Al) max. 500
Calcium (Ca) max. 100
Iron (Fe) max. 100
Potassium (K) max. 100
Magnesium (Mg) max. 100
Sodium (Na) max. 100
Element concentrations are at time of lot release.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR