Ethyl acetate, BAKER ANALYZED® LC-MS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Acetic acid ethyl ester

BAKR9828-03EA 420 SGD
BAKR9828-03
Ethyl acetate, BAKER ANALYZED® LC-MS, J.T.Baker®
Ethyl acetate
Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 - 400 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 265 nm ≤0.050
Ultraviolet Absorbance (1.00-cm cell vs. water) - 255 nm ≤1.00
Assay (CH₃COOC₂H₅) ≥99.6 %
Residue after Evaporation ≤1.0 ppm
Water (H₂O) ≤0.05 %
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤50 ppb
Trace Impurities - Aluminum (Al), For Information Only For Information Only
Trace Impurities - * Calcium (Ca), For Information Only For Information Only
Trace Impurities - Iron (Fe), For Information Only For Information Only
Trace Impurities - Magnesium (Mg), For Information Only For Information Only
Trace Impurities - Potassium (K), For Information Only For Information Only
Trace Impurities - *Sodium (Na), For Information Only
* May change over time due to extraction from glass container.
Filtered through a 0.2 micron filter.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR