Water with 0,1% (v/v) formic acid, BAKER ANALYZED® LC-MS, J.T.Baker®

Supplier: Avantor

BAKR9834-02EA 52.8 SGD
BAKR9834-02 BAKR9834-03
Water with 0,1% (v/v) formic acid, BAKER ANALYZED® LC-MS, J.T.Baker®
Water with 0,1% (v/v) formic acid

Clear, colorless liquid. For use in liquid chromatography and mass spectrometry.

Storage Temperature: Ambient
EINECS: 200-579-1
REACH: 01-2119491174-37-XXXX

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use Passes Test
Appearance Passes Test
Formic Acid 0.095 - 0.105 %
Gradient Elution Test (a.u.) - 254 nm ≤ 0.01
Residue after Evaporation ≤ 1.0 ppm
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤ 50 ppb
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Trace Impurities - * Calcium (Ca) ≤ 50.0 ppb
Trace Impurities - Iron (Fe) ≤ 50.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 50.0 ppb
Trace Impurities - Potassium (K) ≤ 50 ppb
Trace Impurities - * Sodium (Na) ≤ 50.0 ppb
* May change over time due to extraction from glass container.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR