J.T.Baker®, Bovine Serum Albumin (BSA), Fraction V

Supplier: Avantor

BAKRA464-02EA 165 SGD
BAKRA464-02
J.T.Baker®, Bovine Serum Albumin (BSA), Fraction V
Proteins and Peptides

Order Now

SPECIFICATIONS

Protein/peptide name Bovine Serum Albumin
Environmentally Preferable
Protein synonyms BSA|Fraction V|Cohn Fraction V
Species Bovine
Source Serum
CAS No. 9048-46-8
Colour light yellow
Storage conditions Refrigerator

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR