Hydrochloric acid concentrate for dilution to 0.1 mol (0.1N), DILUT-IT® dissolution media concentrate, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

BAKRD011-00EA 0 SGD
BAKRD011-00
Hydrochloric acid concentrate for dilution to 0.1 mol (0.1N), DILUT-IT® dissolution media concentrate, J.T.Baker®
Hydrochloric acid

DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities

Formula: HCl
MW: 36,46 g/mol
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use 0.098 - 0.102
STORE AT ROOM TEMPERATURE
Appearance Passes Test
Density (g/mL) at 25°C, For Information Only
Normality at 26x dilution 0.098 - 0.102
DILUT-IT dissolution media concentrates are produced from raw materials in accordance with USP/EP/BP/JP guidelines under validated processes in Avantor Performance Materials' ISO certified manufacturing facilities

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR