J.T.Baker® DILUT-IT™, Sodium Phosphate, Dissolution Media Concentrate

Supplier: Avantor

BAKRD021-02EA 44.1 SGD
BAKRD021-02
J.T.Baker® DILUT-IT™, Sodium Phosphate, Dissolution Media Concentrate
Buffers Biological Buffers

  • pH at 25 °C at 26X Dilution: 6,75 to 6,85
  • Store at room temperature

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR