1-Butanesulphonic acid sodium salt ≥98%, J.T.Baker®

Supplier: Avantor

BAKRD600-03EA 198 SGD
BAKRD600-03
1-Butanesulphonic acid sodium salt ≥98%, J.T.Baker®
1-Butansulphonic acid sodium salt

Powder.

Formula: C₄H₉NaO₃S
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007540
CAS Number: 2386-54-1
EINECS: 219-201-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use = 98 %
Assay (C₄H₉NaO₃) (by titration) ≥98 %
Appearance Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR