n-Hexane ≥99.0% (by GC), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

Packaged under inert gas.

BAKRN168-08EA 613 SGD
BAKRN168-08
n-Hexane ≥99.0% (by GC), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
n-Hexane

For trace organic analysis by GC and HPLC. UV cutoff 210nm. Purity based on n-hexane and saturated C6 hydrocarbons, corrected for water. Certificate of lot analysis provided.

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69
Melting Pt: –94.3 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use ≥99.5%
ACS - Assay (Total Saturated C₆ Isomers) (by GC, corrected for water) ≥99.5 %
ACS - Assay (as n-Hexane) (by GC, corrected for water) ≥99.0 %
ACS - Color (APHA) ≤10
ACS - Residue after Evaporation ≤3 ppm
ACS - Water-Soluble Titrable Acid, meq/g ≤0.0003
ACS - Sulfur Compounds (as S) ≤0.005 %
ACS - Thiophene Passes Test
ACS - Water (by KF, coulometric) ≤0.03 %
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-280 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 250 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 240 nm ≤0.04
Ultraviolet Absorbance (1.00-cm cell vs. water) - 230 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.20
Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm ≤0.30
Ultraviolet Absorbance (1.00-cm cell vs. water) - 195 nm ≤1.00
Product Information (not specifications):
Density (g/mL) at 25 °C (typical)........ 0.663

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR