Hexane (mixture of isomers) 95 GC, sum of isomers+Methylcyclopentane, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Hexanes, Isohexane

M.O.C. Category N

BAKRN169-08EA 117 SGD
BAKRN169-08
Hexane (mixture of isomers) 95 GC, sum of isomers+Methylcyclopentane, J.T.Baker®
Hexane (mixture of isomers)
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 68…70 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (sum of 5 isomers, total hexanes, plus methylcyclopentane)(by GC) ≥95 %
Identification (by IR) Passes Test
Appearance (Clear, colorless liquid) Passes Test
Color (APHA) ≤10
Residue after Evaporation ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR