4-Hydroxy-4-methyl-2-pentanone, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Diacetone alcohol

Management of Change (MOC) category = N

BAKRN679-09EA 250 SGD
BAKRN679-09
4-Hydroxy-4-methyl-2-pentanone, J.T.Baker®
4-Hydroxy-4-methyl-2-pentanone
Formula: (CH₃)₂C(OH)CH₂COCH₃
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0,94 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004471
CAS Number: 123-42-2
EINECS: 204-626-7
UN: 1148
ADR: 3,III
REACH: 01-2119473975-21
Merck Index: 13,02983

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR