Polyethyleneimine 50% in water, J.T.Baker®

Supplier: Avantor


Management of Change (MOC) category = N

BAKRU230-08EA 832 SGD
BAKRU230-08
Polyethyleneimine 50% in water, J.T.Baker®
Polyethyleneimine
Formula: (CH2CH2NH)n
MDL Number: MFCD00084427
CAS Number: 9002-98-6

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance (colorless to light yellow) Passes Test
Identification (by IR) Passes Test
pH of 5% Solution in water, For Information Only
Residue after Evaporation, For Information Only
Viscosity, For Information Only


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR