Polyvinyl alcohol 87-89%, partially hydrolysed, J.T.Baker®

Supplier: Avantor

Synonyms: Poly(vinyl alcohol), PVOH, PVA, Mowiol®

Management of Change (MOC) category = N

BAKRU232-08EA 203 SGD
BAKRU232-08
Polyvinyl alcohol 87-89%, partially hydrolysed, J.T.Baker®
Polyvinyl alcohol
Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Viscosity of 4% Aqueous Solution at 20°C (cps) 4 - 7
Identification (by IR) Passes Test
pH of 4% Aqueous Solution 4.5 - 6.5
Loss on Drying at 105°C ≤5 %
Residue after Ignition ≤0.75 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR