Ultrapure water system, Barnstead™ GenPure™
Supplier: Thermo Fisher Scientific
50131950
BARN50131950EA 11580 SGD
BARN50131950
Ultrapure water system, Barnstead™ GenPure™
Water Purification Systems Pure and Ultra Pure Water Systems
Suitable for even the most demanding and sensitive applications, the family of Thermo Scientific™ Barnstead™ GenPure™ water purification systems deliver ultrapure 18,2 MΩ.cm water with consistent quality.
  • UV oxidation reduces organic levels to <5 ppb, and protects against microorganisms (only on systems containing a UV lamp (UV))
  • Ultrafiltration removes pyrogens from the feed water
  • Feed water monitoring alerts users to fluctuations in feed water quality
  • Real-time monitor of the amount of organic materials in the product water shown on the display
  • USP-
More Product Information