Bottle/tube rollers
Supplier: Thermo Fisher Scientific
88881004
BARN88881004EA 0 SGD
BARN88881004
Bottle/tube rollers
Shakers Rotating/Rocking Shakers
This sleek compact design unit is capable of rolling multiple size tubes or bottles for applications like cell culture, hybridisation assays and liquid mixing. The anti-corrosive, wear-resistant construction enables use in humid and CO₂ environments.
  • Adjustable speed accommodates a variety of applications
  • Adjustable rollers accommodate a variety of tube/bottle sizes
  • Stacking kit allows two units to be stacked, saving valuable bench space
  • Optional remote control facilitates operating unit while in incubator and adds timer function
Certifications: More Product Information