Orbital shaker, CO₂ resistant
Supplier: Thermo Fisher Scientific
88881102
BARN88881102EA 0 SGD
BARN88881102
Orbital shaker, CO₂ resistant
Shakers Orbital Shakers
This CO₂ resistant orbital shaker provides reliable continuous operation, it is ideally suited to keep cells alive and flourishing within your working environment. The compact shaker can be used both in and outside of a CO₂ incubator, giving maximum flexibility. The external control box features an LED display which shows speed and time, and also has indicator lights for rpm, time, run, plus a visual warning light. Applications include: Mammalian cell culture, bacterial suspensions, staining and destaining, washing procedures, hybridisation, solubility studies, extraction procedures andMore Product Information