Thermo Scientific™ Locator™ Plus Rack and Box Systems
Supplier: Thermo Fisher Scientific
CY50925 CY509109 CY50935 CY50935-70
BARNCY50925EA 9180 SGD
BARNCY50925 BARNCY509109 BARNCY50935 BARNCY50935-70
Thermo Scientific™ Locator™ Plus Rack and Box Systems
Dewars and Cryogenic Vessels
Locator™ cryogenic rack and box systems allow quick, efficient sample retrieval with maximum storage capacity (up to 6000 samples with Locator™ Plus) combined with minimum liquid nitrogen consumption.
  • Compact design minimises space requirements while maximising storage capacity
  • System is suited for both manual and computerised inventory record-keeping methods
  • Super-insulated vessels have special, narrow-mouth design that conserves liquid nitrogen by minimising evaporation
  • Outstanding temperature uniformity: Samples are stored below –180 °C, even when less
More Product Information