VWR® B'Wipes 3000, Cleanroom wipes
Supplier: VWR Collection
BASA124083EA 57.3 SGD
BASA124083
VWR® B'Wipes 3000, Cleanroom wipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Polyester wipe for use in ISO 4 cleanrooms.
  • Single-knit, polyester wipe, with knife-cut edges (130 g/m²)
  • Laundered for better cleanliness
  • Good wiping efficiency and abrasion resistance
  • Ideal for general cleaning of all types of surfaces
  • Available in an economical version for floors