CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
BCS-206B BCS-206 BCS-206G BCS-206O BCS-206P BCS-206PK BCS-207 BCS-207B BCS-207G BCS-207O BCS-207P BCS-207PK BCS-206MC BCS-217G BCS-217P BCS-221G BCS-221P BCS-209G BCS-209P BCS-220G BCS-220P BCS-215G BCS-215P BCS-219G BCS-219P
BCISBCS-206BEA 175 SGD
BCISBCS-206B BCISBCS-206 BCISBCS-206G BCISBCS-206O BCISBCS-206P BCISBCS-206PK BCISBCS-207 BCISBCS-207B BCISBCS-207G BCISBCS-207O BCISBCS-207P BCISBCS-207PK BCISBCS-206MC BCISBCS-217G BCISBCS-217P BCISBCS-221G BCISBCS-221P BCISBCS-209G BCISBCS-209P BCISBCS-220G BCISBCS-220P BCISBCS-215G BCISBCS-215P BCISBCS-219G BCISBCS-219P
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Boxes Cryoboxes
Laminated cardboard.
  • Designed for repeated freezing and thawing in vapor-phase liquid nitrogen
  • Standard boxes available in 6 colours
  • Available in 2 heights, 5,2 cm and 9,2 cm to accommodate 1 to 5 ml cryotubes
TruCool® Cryoboxes have a hinged lid that remains attached which ensures lid is placed on correctly and markings are in sync with samples. Lid is easy to open, even when frozen and stays attached to the base minimising risk of contamination. Boxes have a white patch on the front for markings or barcode. Made from laminated cardboard, waterproofMore Product Information