Environmentally friendly thermometer, individually calibrated, Enviro-Safe®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
B602020100
BELAB602020100EA 943 SGD
BELAB602020100
Environmentally friendly thermometer, individually calibrated, Enviro-Safe®, SP Bel-Art
Thermometers Analogue Thermometers
Each easy to read thermometer includes an individualised thermometer calibration report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • EnviroKleen™ certified
  • Non toxic, Enviro-Safe® liquid, against lead-free, yellow back glass provides easy readability and fewer reading errors