Maximum registering, autoclave bi-metallic thermometers, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
B602150011
BELAB602150011EA 464 SGD
BELAB602150011
Maximum registering, autoclave bi-metallic thermometers, SP Bel-Art
Thermometers Analogue Thermometers
This durable thermometer, with maximum registering pointer (red), is moved up by the temperature measuring pointer (black) to indicate the highest temperature. The threaded connection (¼" NPT) screws into processes and can be used to mount thermometer on included stand.
  • Manual reset
  • Waterproof and pressurised
  • Withstands pressure, high temperatures and rust
  • Calibration point for item 620-2342: 121 °C
  • Dial size: 81 mm
Stainless steel 304 with glass lens.
Delivery information: Supplied with multi-language (English, French, German, Italian, PortugueseMore Product Information