Calibrated electronic verification thermometers, FRIO-Temp®
Supplier: Bel-Art Products, a Part of SP
B610000000
BELAB610000000EA 204 SGD
BELAB610000000
Calibrated electronic verification thermometers, FRIO-Temp®
Thermometers Digital Thermometers
Digital thermometers verify sample temperatures in freezers, refrigerators, incubators and ovens.
  • Accurate temperature reading without sample contamination is achieved by using the filled bottle, which mimics the sample
  • Individually serialised
Ideal for chemical, food/beverage, petrochemistry laboratories, pharmaceutical, medical, cosmetic, university, breweries, beer cellars, refrigerated storage, hot houses, blood banks, refrigeration/freezer/incubator/oven equipment producers and producers of laboratory equipment.
A serial numbered certificateMore Product Information