Safety wash bottles, wide neck, with printed name, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F116460638
BELAF116460638EA 92.3 SGD
BELAF116460638
Safety wash bottles, wide neck, with printed name, SP Bel-Art
Bottles Wash Bottles
LDPE, with PP spray cap.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Wide opening for easy adn safe filling
These bottles include a 53 mm wide PP closure with leakproof design for dispensing most common solvents. Easy-to-read, LDPE solvent-resistant, printed bottles include: Department of Transportation (DOT) and National Fire Protection Association (NFPA) codes and symbols, Chemical Abstract Service (CAS) number, protective equipment symbols and target organ information, making your workplace a safer place. Provides compliance withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable