SP Bel-Art bench-top biohazard bags
Supplier: Bel-Art Products, a Part of SP
F131660001
BELAF131660001EA 661 SGD
BELAF131660001
SP Bel-Art bench-top biohazard bags
Bags Waste Disposal Bags
Bright red, autoclavable bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene.
  • Use as a biohazard material handler: Small size allows the bag to be inverted over your hand to safely pick up waste
  • Steam autoclavable at 121 °C
For a space-saving disposal unit in fume hoods or on bench tops, use with Poxygrid® Benchtop Biohazard Bag Holder (129-4948) and Cover (129-1475).
Certifications: Pass ASTM 1922 tear resistanceMore Product Information