Bench top biohazard bag holder kit, SP Bel-Art Poxygrid®
Supplier: Bel-Art Products, a Part of SP
F131930500
BELAF131930500EA 160 SGD
BELAF131930500
Bench top biohazard bag holder kit, SP Bel-Art Poxygrid®
Racks Bag Racks
Economical and convenient, this bench top disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc during daily use.
  • Snug fitting polypropylene plastic cover helps hold the bag securely in place
  • Reusable, can be cleaned by hand or in automated dish washers
  • Autoclavable
The rectangular steel wire frame holds a 220×280 mm polyethylene bag which is coated in easy to clean, durable, orange colour epoxy.
Certifications: Cover provides compliance with 29 CFR 1910.1030(d)(2)(xiii)(A), which requires a cover on any container usedMore Product Information