Funnels, micro
Supplier: Bel-Art Products, a Part of SP
F146850035
BELAF146850035EA 84.4 SGD
BELAF146850035
Funnels, micro
Funnels Filtration Funnels Filter Funnels
PP
  • Good chemical resistance
  • Autoclavable at 121 °C
Ideal for filling burettes and small bottles.  