Multipurpose trays, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F162000010
BELAF162000010EA 118 SGD
BELAF162000010
Multipurpose trays, SP Bel-Art
Trays/Dishes/Pots Laboratory Trays/Dishes
Trays made of PP.
  • Stackable
  • Top rim adds rigidity and provides handholds for transport
  • Rounded inside corners make cleaning easy
Ideal for holding instruments while autoclaving and cleaning glassware to reduce chance of damage.