Water bath protective agent, Aqua-Clear™, Cleanware™, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F170930000
BELAF170930000EA 0 SGD
BELAF170930000
Water bath protective agent, Aqua-Clear™, Cleanware™, SP Bel-Art
Cleaning Agents Water Bath Protection Reagents
Prevents the growth of algae and bacteria in baths and circulating thermostats, and prevents tubing from clogging.
  • Suppiled as concentrate- use 2 ml per litre of water
Caution: Use biocides safely. Always read the label and product information before use.