Snap cap vials, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F175750005
BELAF175750005EA 170 SGD
BELAF175750005
Snap cap vials, SP Bel-Art
Vials
These snap cap vials are constructed of glass-clear styrene for complete visibility and are excellent for storing dry materials, powders, tablets, and other samples.
  • Heavy, rigid-wall construction ensures long service
  • Can be used at temperatures from –20 to 70 °C
Delivery information: Supplied with tight-fitting, PE snap-on caps.