Tubing clamps, screw style, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F182120000
BELAF182120000EA 90.6 SGD
BELAF182120000
Tubing clamps, screw style, SP Bel-Art
Clamps Tubing and Sanitary Clamps
Molded of Acetal plastic. This tubing clamp offers on/off and fine control so accurate that it replaces stopcocks in many applications.
  • Can be installed without disconnecting tubing or interrupting operation
  • Accepts flexible tubing up to 14 mm Ext.Ø
  • Resistant to acids, alkalis and organic solvents
  • Autoclavable