Holder for Flask immersion weights, ring type, Poxygrid®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F183071000
BELAF183071000EA 78.1 SGD
BELAF183071000
Holder for Flask immersion weights, ring type, Poxygrid®, SP Bel-Art
Weights Flask Weights
This wire formed, epoxy coated holder keeps lead ring weights organised and handy.
  • Small footprint
  • Base Ø: 190 mm
Larger stack of 171 mm height is neatly designed on tapered centre.
Order Now
SPECIFICATIONS
Environmentally Preferable