Wire weights for vessels, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F183240001
BELAF183240001EA 120 SGD
BELAF183240001
Wire weights for vessels, SP Bel-Art
Weights Flask Weights
These lead weights can be coiled into a helix, spiral, cone or any irregular shape to stabilise vessels in a water bath.
  • Flexible and versatile
  • Coil around flasks, thread through handles or loop around bottle necks
  • Coated with chemical resistant red Vikem® vinyl to protect glassware