Rack for cuvettes, No-Wire™, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F185200000
BELAF185200000EA 75.4 SGD
BELAF185200000
Rack for cuvettes, No-Wire™, SP Bel-Art
Racks Cuvette Racks
These PP rack provides gentle no-scratch support for fragile and expensive spectrophotometer cuvettes.
  • Holds forty-two 10 mm light path cuvettes
  • 6×7 row layout
  • Steam autoclavable at 121 °C