Microcentrifuge tube mini floating racks, Prep-Safe™, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F187420120
BELAF187420120EA 31.5 SGD
BELAF187420120
Microcentrifuge tube mini floating racks, Prep-Safe™, SP Bel-Art
Racks Tube Racks
PP, coloured.
  • PP, steam autoclavable at 121 °C
  • Immobilising tabs prevent tube tops from popping open during boiling
  • Holds up to twelve 1,5 or 2,0 ml microcentrifuge tubes in numbered locations
  • Raised handle for easy lifting: stable feet keep the rack upright on the bench
  • 95 mm outer diameter fits most 1000 ml beakers
Unique tabs protect your samples from loss and contamination. Place tubes in slots and slide under the tabs to keep tubes in place and caps closed when under pressure, floating or submerged. For use in water baths, ice or on the benchtop.
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable