Submersible plastic test tube grip racks, Bel-Art No-Wire, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F187490001
BELAF187490001EA 71.9 SGD
BELAF187490001
Submersible plastic test tube grip racks, Bel-Art No-Wire, SP Bel-Art
Racks Tube Racks
Retainer fingers hold tubes firmly in the rack plates with no fear of dislodging, making them perfect for both decanting and water bath work.
  • Fingers allow simultaneous decanting to save valuable time
  • Specially formula plastic allows the rack to submerge in water baths
  • Fingers secure tubes while in water baths to eliminate the risk of diluting or spilling
  • Features end plates for labelling as well as numbered and lettered rows for tube identification
  • Fold up sides lock firmly into the grip decks; assembly takes only seconds
  • Unloaded racks are stackable