Racks for centrifuge tubes, Poxygrid®, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F187940003
BELAF187940003EA 65.1 SGD
BELAF187940003
Racks for centrifuge tubes, Poxygrid®, SP Bel-Art
Racks Tube Racks
Epoxy-coated steel wire, autoclavable.
  • Supports one 50 ml conical centrifuge tube
Hole Ø: 29 mm