Cryo storage boxes, 1 °C Cryo-Safe™, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F188440000
BELAF188440000EA 354 SGD
BELAF188440000
Cryo storage boxes, 1 °C Cryo-Safe™, SP Bel-Art
Boxes Cooling/Freezing Boxes and Bags
Easy-to-use unit provides 'controlled rate' freezing samples at approximately −1 °C per minute, which is needed to prevent lethal intracellular freezing and successful cryopreservation. Prior to inserting vials, the unit should be filled with 250 ml of 100% isopropyl alcohol. Once the samples have been inserted the unit should be placed in a −80 °C mechanical freezer.
  • Clear polycarbonate container body; high-density polyethylene screw closure (Ø 124 mm)
  • 18-well, removable polyethylene vial holder, placed inside the container, separates the vials from the
More Product Information