Cryo storage box, 0 °C Cryo-Safe™ Junior Cooler, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
F188461000
BELAF188461000EA 1160 SGD
BELAF188461000
Cryo storage box, 0 °C Cryo-Safe™ Junior Cooler, SP Bel-Art
Boxes Cooling/Freezing Boxes and Bags
This benchtop sized cooler with polycarbonate body and clear polycarbonate lid, holds twelve 15 ml centrifuge or cryo vials at a temperature below 0 °C for approximately 3 hours. Store at –20 °C for a few hours until the temperature of the cooler falls below 0 °C.
  • Tubes up to 125 mm tall can be accommodated when the lid is secured closed with the built-in wire carrying handle
  • Cooler body is filled with a non-toxic gel which cannot contact tubes; ensures markings and labels on tubes are safe
  • Corresponding tube positions are printed on the keyed lid and
More Product Information